Assay Details
| Gene Symbol: akt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029441.4 | NP_001024612.1 | 6 - 7 | 928 | 111 | |
| NM_077956.8 | NP_510357.3 | 6 - 7 | 928 | 111 | ||
| GenBank mRNA | AF072381.1 | - | 6 - 7 | 928 | 111 | |
More Information
Set Membership: |
|

akt-2
Genomic Map