Assay Details
| Gene Symbol: del-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368544.2 | NP_001355359.1 | 4 - 5 | 497 | 82 | |
| NM_001368545.3 | NP_001355470.1 | 5 | 641 | 82 | ||
| NM_001368546.2 | NP_001355471.1 | 4 - 5 | 446 | 82 | ||
More Information
Set Membership: |
|

del-1
Genomic Map