Assay Details
| Gene Symbol: del-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368544.2 | NP_001355359.1 | 8 - 9 | 849 | 95 | |
| NM_001368545.3 | NP_001355470.1 | 9 | 993 | 95 | ||
| NM_001368546.2 | NP_001355471.1 | 8 - 9 | 798 | 95 | ||
More Information
Set Membership: |
|

del-1
Genomic Map