Assay Details
| Gene Symbol: prx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077985.6 | NP_510386.2 | 15 - 16 | 2154 | 91 | |
| GenBank mRNA | AB054992.1 | - | 15 - 16 | 2154 | 91 | |
More Information
Set Membership: |
|

prx-1
Genomic Map