Assay Details
| Gene Symbol: prx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381145.1 | NP_001367171.1 | 3 - 4 | 270 | 83 | |
| NM_077985.6 | NP_510386.2 | 20 - 21 | 2669 | 83 | ||
| GenBank mRNA | AB054992.1 | - | 20 - 21 | 2669 | 83 | |
More Information
Set Membership: |
|

prx-1
Genomic Map