Assay Details
| Gene Symbol: prx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077985.6 | NP_510386.2 | 2 - 3 | 489 | 90 | |
| GenBank mRNA | AB054992.1 | - | 2 - 3 | 489 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prx-1| Gene Symbol: prx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077985.6 | NP_510386.2 | 2 - 3 | 489 | 90 | |
| GenBank mRNA | AB054992.1 | - | 2 - 3 | 489 | 90 | |
Set Membership: |
|