Assay Details
| Gene Symbol: wdr-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077993.5 | NP_510394.1 | 1 - 2 | 125 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-5.2| Gene Symbol: wdr-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077993.5 | NP_510394.1 | 1 - 2 | 125 | 103 | |
Set Membership: |
|