Assay Details
| Gene Symbol: ife-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268267.4 | NP_001255196.1 | 3 - 4 | 1336 | 132 | |
| NM_001268268.2 | NP_001255197.1 | 1 - 2 | 465 | 132 | ||
| Gene Symbol: dro-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379982.1 | NP_001366888.1 | 3 - 4 | 1362 | 132 | |
More Information
Set Membership: |
|

ife-1
Genomic Map