Assay Details
| Gene Symbol: skr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074658.6 | NP_507059.1 | 3 - 4 | 439 | 170 | |
| GenBank mRNA | AF440507.1 | - | 2 - 3 | 384 | 170 | |
More Information
Set Membership: |
|

skr-3
Genomic Map