Assay Details
| Gene Symbol: cin-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063039.7 | NP_495440.4 | 2 - 3 | 248 | 138 | |
| GenBank mRNA | EU191083.1 | - | 2 - 3 | 246 | 138 | |
More Information
Set Membership: |
|

cin-4
Genomic Map