Assay Details
| Gene Symbol: clpr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060502.2 | NP_492903.2 | 6 | 875 | 114 | |
| Gene Symbol: clpr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060503.2 | NP_492904.2 | 6 | 875 | 114 | |
More Information
Set Membership: |
|

clpr-4
Genomic Map