Assay Details
Gene Symbol: fbxb-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064755.5 | NP_497156.1 | 3 | 1016 | 120 |
Gene Symbol: fbxb-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064754.3 | NP_497155.1 | 2 | 529 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_064754.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
