Assay Details
| Gene Symbol: F26F4.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065630.6 | NP_498031.3 | 2 | 208 | 125 | |
| NM_065632.8 | NP_498033.1 | 2 | 166 | 125 | ||
| Gene Symbol: F26F4.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065629.5 | NP_498030.1 | 2 | 188 | 125 | |
| Gene Symbol: cee-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065633.10 | NP_498034.1 | 1 | 47 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_065629.5, NM_065633.10 transcripts.
Set Membership: |
|

F26F4.9
Genomic Map