Assay Details
| Gene Symbol: nlp-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372660.1 | NP_001359616.1 | 2 | 481 | 67 | |
| NM_067065.6 | NP_499466.1 | 2 | 479 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nlp-21| Gene Symbol: nlp-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372660.1 | NP_001359616.1 | 2 | 481 | 67 | |
| NM_067065.6 | NP_499466.1 | 2 | 479 | 67 | ||
Set Membership: |
|