Assay Details
| Gene Symbol: mtl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072295.6 | NP_504696.1 | 2 | 252 | 105 | |
| GenBank mRNA | X53244.1 | - | 237 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtl-1| Gene Symbol: mtl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072295.6 | NP_504696.1 | 2 | 252 | 105 | |
| GenBank mRNA | X53244.1 | - | 237 | 105 | ||
Set Membership: |
|