Assay Details
| Gene Symbol: srab-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028376.2 | NP_001023547.1 | 4 - 5 | 542 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srab-24| Gene Symbol: srab-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028376.2 | NP_001023547.1 | 4 - 5 | 542 | 108 | |
Set Membership: |
|