Assay Details
| Gene Symbol: ZK381.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028443.3 | NP_001023614.2 | 3 - 4 | 583 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZK381.8| Gene Symbol: ZK381.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028443.3 | NP_001023614.2 | 3 - 4 | 583 | 109 | |
Set Membership: |
|