Assay Details
| Gene Symbol: C13C4.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028485.6 | NP_001023656.2 | 3 - 4 | 627 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C13C4.7| Gene Symbol: C13C4.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028485.6 | NP_001023656.2 | 3 - 4 | 627 | 109 | |
Set Membership: |
|