Assay Details
| Gene Symbol: clc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029822.6 | NP_001024993.1 | 6 - 7 | 717 | 95 | |
| Gene Symbol: sorf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076183.9 | NP_508584.3 | 6 - 7 | 716 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001029822.6 transcripts.
Set Membership: |
|

clc-3
Genomic Map