Assay Details
| Gene Symbol: sra-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063945.6 | NP_496346.2 | 3 - 4 | 491 | 118 | |
| Gene Symbol: gly-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063944.7 | NP_496345.1 | 4 - 5 | 544 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063944.7 transcripts.
Set Membership: |
|

sra-12
Genomic Map