Assay Details
| Gene Symbol: srx-113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073757.2 | NP_506158.2 | 3 - 4 | 842 | 118 | |
| Gene Symbol: F56H9.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383423.1 | 4 - 5 | 783 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383423.1 transcripts.
Set Membership: |
|

srx-113
Genomic Map