Assay Details
| Gene Symbol: C33E10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078404.4 | NP_510805.2 | 1 - 2 | 389 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C33E10.8| Gene Symbol: C33E10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078404.4 | NP_510805.2 | 1 - 2 | 389 | 140 | |
Set Membership: |
|