Assay Details
| Gene Symbol: mbd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025841.4 | NP_001021012.1 | 1 - 2 | 190 | 58 | |
| GenBank mRNA | DQ010405.1 | - | 1 - 2 | 212 | 58 | |
More Information
Set Membership: |
|

mbd-2
Genomic Map