Assay Details
| Gene Symbol: mbd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025841.4 | NP_001021012.1 | 2 - 3 | 307 | 117 | |
| GenBank mRNA | DQ010405.1 | - | 2 - 3 | 329 | 117 | |
More Information
Set Membership: |
|

mbd-2
Genomic Map