Assay Details
| Gene Symbol: srz-83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026143.1 | NP_001021314.1 | 3 - 4 | 555 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srz-83| Gene Symbol: srz-83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026143.1 | NP_001021314.1 | 3 - 4 | 555 | 109 | |
Set Membership: |
|