Assay Details
| Gene Symbol: F56G4.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026339.2 | NP_001021510.1 | 3 - 4 | 267 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F56G4.7| Gene Symbol: F56G4.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026339.2 | NP_001021510.1 | 3 - 4 | 267 | 73 | |
Set Membership: |
|