Assay Details
| Gene Symbol: aqp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264219.3 | NP_001251148.1 | 6 - 7 | 646 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aqp-9| Gene Symbol: aqp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264219.3 | NP_001251148.1 | 6 - 7 | 646 | 116 | |
Set Membership: |
|