Assay Details
| Gene Symbol: aqp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264219.3 | NP_001251148.1 | 7 - 8 | 782 | 107 | |
| NM_001264220.3 | NP_001251149.1 | 1 - 2 | 109 | 107 | ||
More Information
Set Membership: |
|

aqp-9
Genomic Map