Assay Details
| Gene Symbol: Y6B3B.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377673.1 | NP_001364617.1 | 1 - 2 | 74 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y6B3B.4| Gene Symbol: Y6B3B.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377673.1 | NP_001364617.1 | 1 - 2 | 74 | 77 | |
Set Membership: |
|