Assay Details
| Gene Symbol: srz-67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026794.1 | NP_001021965.1 | 1 - 2 | 160 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srz-67| Gene Symbol: srz-67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026794.1 | NP_001021965.1 | 1 - 2 | 160 | 148 | |
Set Membership: |
|