Assay Details
| Gene Symbol: stg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026805.2 | NP_001021976.1 | 3 - 4 | 425 | 70 | |
| GenBank mRNA | DQ015968.1 | - | 3 - 4 | 347 | 70 | |
More Information
Set Membership: |
|

stg-1
Genomic Map