Assay Details
| Gene Symbol: ins-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026982.3 | NP_001022153.1 | 3 - 4 | 367 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ins-14| Gene Symbol: ins-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026982.3 | NP_001022153.1 | 3 - 4 | 367 | 102 | |
Set Membership: |
|