Assay Details
| Gene Symbol: fut-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027139.2 | NP_001022310.1 | 5 - 6 | 804 | 82 | |
| GenBank mRNA | AJ745073.1 | - | 5 - 6 | 720 | 82 | |
More Information
Set Membership: |
|

fut-5
Genomic Map