Assay Details
Gene Symbol: aqp-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027309.3 | NP_001022480.1 | 3 - 4 | 510 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
