Assay Details
| Gene Symbol: ins-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027358.5 | NP_001022529.2 | 1 - 2 | 259 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ins-20| Gene Symbol: ins-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027358.5 | NP_001022529.2 | 1 - 2 | 259 | 153 | |
Set Membership: |
|