Assay Details
| Gene Symbol: ins-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381761.1 | NP_001367970.1 | 1 - 2 | 140 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ins-16| Gene Symbol: ins-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381761.1 | NP_001367970.1 | 1 - 2 | 140 | 107 | |
Set Membership: |
|