Assay Details
| Gene Symbol: grl-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027854.5 | NP_001023025.1 | 2 - 3 | 709 | 64 | |
| NR_101843.1 | 2 - 3 | 707 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

grl-25| Gene Symbol: grl-25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027854.5 | NP_001023025.1 | 2 - 3 | 709 | 64 | |
| NR_101843.1 | 2 - 3 | 707 | 64 | |||
Set Membership: |
|