Assay Details
| Gene Symbol: mpz-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028170.3 | NP_001023341.2 | 5 - 6 | 512 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mpz-5| Gene Symbol: mpz-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028170.3 | NP_001023341.2 | 5 - 6 | 512 | 106 | |
Set Membership: |
|