Assay Details
| Gene Symbol: sri-78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062066.2 | NP_494467.2 | 4 - 5 | 804 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-78| Gene Symbol: sri-78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062066.2 | NP_494467.2 | 4 - 5 | 804 | 134 | |
Set Membership: |
|