Assay Details
| Gene Symbol: gsp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027445.6 | NP_001022616.1 | 4 - 5 | 669 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gsp-2| Gene Symbol: gsp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027445.6 | NP_001022616.1 | 4 - 5 | 669 | 74 | |
Set Membership: |
|