Assay Details
| Gene Symbol: tbcc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038294.4 | NP_001033383.3 | 5 - 6 | 647 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tbcc-1| Gene Symbol: tbcc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038294.4 | NP_001033383.3 | 5 - 6 | 647 | 92 | |
Set Membership: |
|