Assay Details
| Gene Symbol: srz-105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038331.2 | NP_001033420.1 | 2 - 3 | 554 | 120 | |
| Gene Symbol: srz-104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028097.3 | NP_001023268.2 | 6 - 7 | 1405 | 120 | |
More Information
Set Membership: |
|

srz-105
Genomic Map