Assay Details
| Gene Symbol: ntc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038459.2 | NP_001033548.1 | 2 - 3 | 265 | 80 | |
| GenBank mRNA | JQ277480.1 | - | 2 - 3 | 229 | 80 | |
More Information
Set Membership: |
|

ntc-1
Genomic Map