Assay Details
| Gene Symbol: W08F4.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061364.3 | NP_493765.3 | 1 - 2 | 90 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

W08F4.10| Gene Symbol: W08F4.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061364.3 | NP_493765.3 | 1 - 2 | 90 | 69 | |
Set Membership: |
|