Assay Details
| Gene Symbol: sri-57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061927.4 | NP_494328.2 | 5 - 6 | 1231 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-57| Gene Symbol: sri-57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061927.4 | NP_494328.2 | 5 - 6 | 1231 | 111 | |
Set Membership: |
|