Assay Details
| Gene Symbol: bnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047660.4 | NP_001041125.1 | 1 - 2 | 120 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bnc-1| Gene Symbol: bnc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047660.4 | NP_001041125.1 | 1 - 2 | 120 | 91 | |
Set Membership: |
|