Assay Details
| Gene Symbol: mpst-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073841.4 | NP_506242.2 | 5 - 6 | 855 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mpst-7| Gene Symbol: mpst-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073841.4 | NP_506242.2 | 5 - 6 | 855 | 118 | |
Set Membership: |
|