Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110822.2 | XP_013966297.1 | 8 - 9 | 1191 | 64 | |
| XM_014110823.2 | XP_013966298.1 | 7 - 8 | 1150 | 64 | ||
| XM_022414902.1 | XP_022270610.1 | 8 - 9 | 1279 | 64 | ||
| XM_022414903.1 | XP_022270611.1 | 6 - 7 | 1132 | 64 | ||
| XM_022414904.1 | XP_022270612.1 | 6 - 7 | 739 | 64 | ||
| XM_022414905.1 | XP_022270613.1 | 6 - 7 | 909 | 64 | ||
| XM_022414906.1 | XP_022270614.1 | 6 - 7 | 844 | 64 | ||
| XM_022414908.1 | XP_022270616.1 | 6 - 7 | 894 | 64 | ||
| XM_022414909.1 | XP_022270617.1 | 6 - 7 | 981 | 64 | ||
| XM_022414910.1 | XP_022270618.1 | 6 - 7 | 907 | 64 | ||
| XM_022414911.1 | XP_022270619.1 | 7 - 8 | 766 | 64 | ||
| XM_022414912.1 | XP_022270620.1 | 6 - 7 | 784 | 64 | ||
| XM_022414913.1 | XP_022270621.1 | 6 - 7 | 756 | 64 | ||
| XM_022414914.1 | XP_022270622.1 | 6 - 7 | 886 | 64 | ||
| XM_022414915.1 | XP_022270623.1 | 6 - 7 | 726 | 64 | ||
| XM_022414916.1 | XP_022270624.1 | 6 - 7 | 777 | 64 | ||
| XM_022414917.1 | XP_022270625.1 | 6 - 7 | 807 | 64 | ||
| XM_022414918.1 | XP_022270626.1 | 8 - 9 | 1187 | 64 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF385B
Genomic Map