Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630263.2 | XP_005630320.1 | 10 - 11 | 1548 | 77 | |
| XM_532910.6 | XP_532910.2 | 9 - 10 | 1625 | 77 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPM1G
Genomic Map