Assay Details
| Gene Symbol: BCL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117544.2 | XP_013973019.1 | 2 - 3 | 530 | 152 | |
| XM_022424767.1 | XP_022280475.1 | 2 - 3 | 483 | 152 | ||
| XM_022424768.1 | XP_022280476.1 | 2 - 3 | 444 | 152 | ||
| XM_860443.5 | XP_865536.3 | 2 - 3 | 402 | 152 | ||
More Information
Set Membership: |
|

BCL11A
Genomic Map